Assay Details
| Gene Symbol: Amica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766180.1 | XP_008764402.1 | 2 - 3 | 189 | 84 | |
| XM_236198.8 | XP_236198.6 | 4 - 5 | 452 | 84 | ||
More Information
Set Membership: |
|

Amica1
Genomic Map