Assay Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022387.3 | NP_071782.1 | 2 - 3 | 201 | 117 | |
| XM_006242957.2 | XP_006243019.1 | 2 - 3 | 667 | 117 | ||
| XM_006242958.2 | XP_006243020.1 | 2 - 3 | 200 | 117 | ||
| XM_008766172.1 | XP_008764394.1 | 2 - 3 | 431 | 117 | ||
| GenBank mRNA | AF016048.1 | - | 2 - 3 | 164 | 117 | |
| BC081714.1 | - | 2 - 3 | 201 | 7108923 | 117 | |
| FQ212439.1 | - | 2 - 3 | 489 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Pafah1b2
Genomic Map