Assay Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022387.3 | NP_071782.1 | 1 - 2 | 117 | 100 | |
| GenBank mRNA | BC081714.1 | - | 1 - 2 | 117 | 7108923 | 100 |
| FQ212439.1 | - | 1 - 2 | 405 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC081714.1,NM_022387.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pafah1b2
Genomic Map