Assay Details
| Gene Symbol: Ttc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005546.1 | NP_001005546.1 | 1 - 2 | 258 | 82 | |
| GenBank mRNA | BC082024.1 | - | 1 - 2 | 258 | 7129913 | 82 |
| FQ210191.1 | - | 1 - 2 | 206 | 82 | ||
| FQ211414.1 | - | 1 - 2 | 206 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc36
Genomic Map