Assay Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013168.2 | NP_037300.2 | 11 - 12 | 907 | 77 | |
| XM_006242896.1 | XP_006242958.1 | 11 - 12 | 993 | 77 | ||
| XM_006242897.1 | XP_006242959.1 | 10 - 11 | 915 | 77 | ||
| XM_008766148.1 | XP_008764370.1 | 10 - 11 | 1077 | 77 | ||
| XM_008766149.1 | XP_008764371.1 | 10 - 11 | 1014 | 77 | ||
| GenBank mRNA | BC088162.1 | - | 11 - 12 | 907 | 7375228 | 77 |
| FQ225972.1 | - | 5 - 6 | 392 | 77 | ||
| FQ226062.1 | - | 11 - 12 | 830 | 77 | ||
| FQ230962.1 | - | 5 - 6 | 392 | 77 | ||
| FQ234793.1 | - | 3 - 4 | 279 | 77 | ||
| FQ234870.1 | - | 9 - 10 | 1063 | 77 | ||
| Y12006.1 | - | 11 - 12 | 777 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmbs
Genomic Map