Assay Details
| Gene Symbol: Il18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY077842.1 | - | 74 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY077842.1 transcripts.
Set Membership: |
|

Il18
Genomic Map