Assay Details
| Gene Symbol: RGD1305464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025011.2 | NP_001020182.2 | 1 - 2 | 179 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025011.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1305464
Genomic Map