Assay Details
| Gene Symbol: Rasl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108162.1 | NP_001101632.1 | 1 - 2 | 202 | 87 | |
| XM_006243260.2 | XP_006243322.1 | 1 - 2 | 210 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rasl12
Genomic Map