Assay Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107168.1 | NP_001100638.1 | 1 - 2 | 50 | 62 | |
| XM_008769454.1 | XP_008767676.1 | 1 - 2 | 50 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107168.1, XM_008769454.1 transcripts.
Set Membership: |
|

Serpinb13
Genomic Map