Assay Details
| Gene Symbol: Fsd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763722.1 | XP_008761944.1 | 12 - 13 | 1493 | 81 | |
| XM_008763723.1 | XP_008761945.1 | 11 - 12 | 1459 | 81 | ||
| XM_008763724.1 | XP_008761946.1 | 11 - 12 | 1457 | 81 | ||
| XM_008763725.1 | XP_008761947.1 | 11 - 12 | 1392 | 81 | ||
More Information
Set Membership: |
|

Fsd1l
Genomic Map