Assay Details
| Gene Symbol: Sall4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749632.3 | XP_003749680.1 | 1 - 2 | 351 | 105 | |
| XM_006235657.2 | XP_006235719.1 | 1 - 2 | 351 | 105 | ||
More Information
Set Membership: |
|

Sall4
Genomic Map