Assay Details
| Gene Symbol: Sall4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749632.3 | XP_003749680.1 | 2 - 3 | 2713 | 96 | |
| XM_006235656.2 | XP_006235718.1 | 2 - 3 | 3178 | 96 | ||
| XM_006235657.2 | XP_006235719.1 | 3 - 4 | 2074 | 96 | ||
More Information
Set Membership: |
|

Sall4
Genomic Map