Assay Details
| Gene Symbol: Tmem189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113752.1 | NP_001107224.1 | 1 - 2 | 260 | 65 | |
| GenBank mRNA | BC158660.1 | - | 1 - 2 | 260 | 8369614 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem189
Genomic Map