Assay Details
| Gene Symbol: Actr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108600.1 | NP_001102070.1 | 3 - 4 | 1012 | 64 | |
| XM_006235422.2 | XP_006235484.1 | 3 - 4 | 1033 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actr5
Genomic Map