Assay Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145670.4 | NP_663703.3 | 9 - 10 | 1413 | 60 | |
| XM_008762466.1 | XP_008760688.1 | 10 - 11 | 1548 | 60 | ||
| GenBank mRNA | AB029897.1 | - | 7 - 8 | 888 | 60 | |
| AB180269.1 | - | 8 - 9 | 1056 | 60 | ||
| AB180272.1 | - | 6 - 7 | 753 | 60 | ||
| BC103480.1 | - | 8 - 9 | 1311 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P039801 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P043886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80016 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039801 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043886 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039801 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043886 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P043886 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P039801 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P198474 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1041880 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16031 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22453 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7832 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88701 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90768 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10852058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852061 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852068 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852069 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852065 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852059 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852062 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852067 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852064 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852070 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852063 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852066 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10852056 | - | - |
| Rat Genome 230 2.0 Array | 1373098_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373098_at | - | - |
| Affymetrix HT Rat Focus Array | 1373098_at | - | - |
| Affymetrix HT Rat Focus Array | 1373098_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300037446.rn.1 | TC0300004899.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300037444.rn.1 | TC0300004899.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300037445.rn.1 | TC0300004899.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373098_at | - | - |
| Rat Expression Set 230 | 1373098_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852056 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22273170 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354404 | - | - |
| RatRef-12 Expression BeadChip | ILMN_55620 | - | - |

Bcas1
Genomic Map