Assay Details
| Gene Symbol: Tp53tg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749631.3 | XP_003749679.2 | 3 - 4 | 918 | 90 | |
| XM_008762554.1 | XP_008760776.1 | 3 - 4 | 918 | 90 | ||
| XM_008762555.1 | XP_008760777.1 | 3 - 4 | 892 | 90 | ||
More Information
Set Membership: |
|

Tp53tg5
Genomic Map