Assay Details
| Gene Symbol: Mroh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235390.2 | XP_006235452.1 | 2 - 3 | 2162 | 62 | |
| XM_008762424.1 | XP_008760646.1 | 2 - 3 | 2162 | 62 | ||
| XM_008762425.1 | XP_008760647.1 | 2 - 3 | 2163 | 62 | ||
| XM_008762426.1 | XP_008760648.1 | 2 - 3 | 2163 | 62 | ||
| XM_008762427.1 | XP_008760649.1 | 2 - 3 | 2163 | 62 | ||
More Information
Set Membership: |
|

Mroh8
Genomic Map