Assay Details
| Gene Symbol: LOC499179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759598.1 | XP_008757820.1 | 3 | 195 | 76 | |
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022615.1 | NP_072137.1 | 2 - 3 | 57 | 76 | |
| GenBank mRNA | AF140782.1 | - | 2 - 3 | 57 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC499179
Genomic Map