Assay Details
| Gene Symbol: Srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014185.2 | NP_001014207.2 | 3 - 4 | 511 | 79 | |
| GenBank mRNA | BC087121.1 | - | 3 - 4 | 502 | 7106749 | 79 |
| FM060375.1 | - | 3 - 4 | 511 | 79 | ||
| FM081417.1 | - | 1 - 2 | 35 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Srsf6
Genomic Map