Assay Details
| Gene Symbol: Zfp335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762559.1 | XP_008760781.1 | 16 - 17 | 3135 | 93 | |
| XM_008762560.1 | XP_008760782.1 | 17 - 18 | 3809 | 93 | ||
| XM_008762561.1 | XP_008760783.1 | 16 - 17 | 3132 | 93 | ||
| XM_008762562.1 | XP_008760784.1 | 16 - 17 | 3135 | 93 | ||
| XM_008762563.1 | XP_008760785.1 | 16 - 17 | 3135 | 93 | ||
| XM_008762564.1 | XP_008760786.1 | 16 - 17 | 3132 | 93 | ||
| XM_008762565.1 | XP_008760787.1 | 16 - 17 | 3135 | 93 | ||
| GenBank mRNA | AF309071.1 | - | 2 - 3 | 157 | 93 | |
| AY079168.1 | - | 8 - 9 | 907 | 93 | ||
More Information
Set Membership: |
|

Zfp335
Genomic Map