Assay Details
| Gene Symbol: Wfdc15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235606.2 | XP_006235668.1 | 3 - 4 | 864 | 77 | |
| XM_006235607.2 | XP_006235669.1 | 2 - 3 | 1129 | 77 | ||
More Information
Set Membership: |
|

Wfdc15b
Genomic Map