Assay Details
| Gene Symbol: LOC102556339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257127.2 | XP_006257189.1 | 4 - 5 | 755 | 67 | |
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191723.1 | NP_001178652.1 | 12 - 13 | 2015 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102556339
Genomic Map