Assay Details
| Gene Symbol: LOC100910121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750759.3 | XP_003750807.1 | 1 - 2 | 271 | 97 | |
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135846.1 | NP_001129318.1 | 16 - 17 | 1722 | 97 | |
| XM_008767458.1 | XP_008765680.1 | 16 - 17 | 1746 | 97 | ||
| XM_008767459.1 | XP_008765681.1 | 16 - 17 | 2097 | 97 | ||
| GenBank mRNA | BC166485.1 | - | 16 - 17 | 1722 | 7099831 | 97 |
| FQ228986.1 | - | 16 - 17 | 1742 | 97 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910121
Genomic Map