Assay Details
| Gene Symbol: LOC103690022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767476.1 | XP_008765698.1 | 1 - 2 | 165 | 101 | |
| Gene Symbol: Ntan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025124.1 | NP_001020295.1 | 109 | 101 | ||
| GenBank mRNA | BC091175.1 | - | 109 | 7321868 | 101 | |
| FQ230577.1 | - | 105 | 101 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690022
Genomic Map