Assay Details
| Gene Symbol: LOC103690022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767472.1 | XP_008765694.1 | 1 - 2 | 179 | 96 | |
| XM_008767473.1 | XP_008765695.1 | 1 - 2 | 179 | 96 | ||
| XM_008767474.1 | XP_008765696.1 | 1 - 2 | 179 | 96 | ||
| XM_008767475.1 | XP_008765697.1 | 1 - 2 | 179 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690022
Genomic Map