Assay Details
| Gene Symbol: Sept12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100865.1 | NP_001094335.1 | 8 - 9 | 892 | 57 | |
| GenBank mRNA | BC097401.1 | - | 8 - 9 | 892 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept12
Genomic Map