Assay Details
| Gene Symbol: Smim22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108266.1 | NP_001101736.1 | 1 - 2 | 104 | 94 | |
| XM_006245806.2 | XP_006245868.1 | 1 - 2 | 752 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108266.1, XM_006245806.2 transcripts.
Set Membership: |
|

Smim22
Genomic Map