Assay Details
| Gene Symbol: Smim22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108266.1 | NP_001101736.1 | 3 - 4 | 355 | 84 | |
| XM_006245805.2 | XP_006245867.1 | 3 - 4 | 920 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smim22
Genomic Map