Assay Details
| Gene Symbol: Tvp23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108263.1 | NP_001101733.1 | 5 - 6 | 937 | 70 | |
| XM_006245717.2 | XP_006245779.1 | 5 - 6 | 1105 | 70 | ||
| XM_006245724.2 | XP_006245786.1 | 5 - 6 | 1102 | 70 | ||
| XM_006245725.2 | XP_006245787.1 | 6 - 7 | 853 | 70 | ||
| XM_006245726.2 | XP_006245788.1 | 6 - 7 | 849 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tvp23a
Genomic Map