Assay Details
| Gene Symbol: Tvp23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108263.1 | NP_001101733.1 | 6 - 7 | 997 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P046503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155726 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046503 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P046503 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P046503 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P220584 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30515 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79937 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10731523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731519 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731518 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731520 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10731517 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000000725.rn.1 | TC1000000075.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10731517 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21879555 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366327 | - | - |

Tvp23a
Genomic Map