Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191588.1 | NP_001178517.1 | 4 - 5 | 544 | 110 | |
| XM_006245784.1 | XP_006245846.1 | 3 - 4 | 823 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nmral1
Genomic Map