Assay Details
| Gene Symbol: Alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108264.1 | NP_001101734.1 | 6 - 7 | 745 | 120 | |
| XM_006245802.2 | XP_006245864.1 | 6 - 7 | 781 | 120 | ||
| XM_008767491.1 | XP_008765713.1 | 6 - 7 | 587 | 120 | ||
| XM_008767492.1 | XP_008765714.1 | 5 - 6 | 1645 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alg1
Genomic Map