Assay Details
| Gene Symbol: Nudt16l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100782.1 | NP_001094252.1 | 1 - 2 | 169 | 83 | |
| GenBank mRNA | BC105819.1 | - | 1 - 2 | 109 | 83 | |
| BC158814.1 | - | 1 - 2 | 169 | 7371601 | 83 | |
| FQ213246.1 | - | 1 - 2 | 159 | 83 | ||
| FQ214509.1 | - | 1 - 2 | 178 | 83 | ||
| FQ219409.1 | - | 1 - 2 | 164 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nudt16l1
Genomic Map