Assay Details
| Gene Symbol: Nudt16l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100782.1 | NP_001094252.1 | 2 - 3 | 429 | 131 | |
| GenBank mRNA | BC105819.1 | - | 2 - 3 | 369 | 131 | |
| BC158814.1 | - | 2 - 3 | 429 | 7371601 | 131 | |
| FQ213246.1 | - | 2 - 3 | 419 | 131 | ||
| FQ214509.1 | - | 2 - 3 | 438 | 131 | ||
| FQ219409.1 | - | 2 - 3 | 424 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nudt16l1
Genomic Map