Assay Details
| Gene Symbol: Mmrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001071128.5 | XP_001071128.2 | 5 - 6 | 1247 | 69 | |
| XM_006236596.2 | XP_006236658.1 | 5 - 6 | 1247 | 69 | ||
| XM_006236597.2 | XP_006236659.1 | 6 - 7 | 1245 | 69 | ||
More Information
Set Membership: |
|

Mmrn1
Genomic Map