Assay Details
| Gene Symbol: Reg3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145846.2 | NP_001139318.1 | 3 - 4 | 248 | 89 | |
| NM_172077.2 | NP_742074.2 | 3 - 4 | 350 | 89 | ||
| GenBank mRNA | CF249035.1 | - | 2 - 3 | 237 | 89 | |
| D23676.1 | - | 3 - 4 | 248 | 89 | ||
| L10229.1 | - | 3 - 4 | 350 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Reg3a
Genomic Map