Assay Details
| Gene Symbol: Pole4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108634.1 | NP_001102104.1 | 1 - 2 | 283 | 143 | |
| GenBank mRNA | FQ213769.1 | - | 1 - 2 | 257 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pole4
Genomic Map