Assay Details
| Gene Symbol: Cnbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022598.1 | NP_072120.1 | 3 - 4 | 385 | 86 | |
| XM_006236854.2 | XP_006236916.1 | 3 - 4 | 380 | 86 | ||
| GenBank mRNA | AB158421.1 | - | 2 - 3 | 220 | 86 | |
| AB158422.1 | - | 2 - 3 | 220 | 86 | ||
| AF242550.1 | - | 3 - 4 | 271 | 86 | ||
| BC062225.1 | - | 3 - 4 | 369 | 6889682 | 86 | |
| D45254.1 | - | 3 - 4 | 385 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cnbp
Genomic Map