Assay Details
| Gene Symbol: Prickle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107876.1 | NP_001101346.1 | 5 - 6 | 724 | 93 | |
| XM_006236912.2 | XP_006236974.1 | 5 - 6 | 1363 | 93 | ||
| XM_006236913.2 | XP_006236975.1 | 5 - 6 | 5673 | 93 | ||
| XM_008763124.1 | XP_008761346.1 | 5 - 6 | 1364 | 93 | ||
| XM_008763125.1 | XP_008761347.1 | 5 - 6 | 702 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prickle2
Genomic Map