Assay Details
| Gene Symbol: Mrpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106596.1 | NP_001100066.1 | 1 - 2 | 86 | 78 | |
| XM_006236635.1 | XP_006236697.1 | 1 - 2 | 43 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006236635.1 transcripts.
Set Membership: |
|

Mrpl35
Genomic Map