Assay Details
| Gene Symbol: Rab43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024331.1 | NP_001019502.1 | 1 - 2 | 302 | 65 | |
| GenBank mRNA | AY593256.1 | - | 1 - 2 | 302 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab43
Genomic Map