Assay Details
| Gene Symbol: Shq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134713.1 | NP_001128185.1 | 237 | 111 | ||
| XM_008763162.1 | XP_008761384.1 | 245 | 111 | |||
| XM_008763164.1 | XP_008761386.1 | 1 - 2 | 245 | 111 | ||
| GenBank mRNA | BC168172.1 | - | 237 | 9026449 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shq1
Genomic Map