Assay Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236885.2 | XP_006236947.1 | 10 - 11 | 1915 | 118 | |
| XM_006236886.2 | XP_006236948.1 | 9 - 10 | 1739 | 118 | ||
More Information
Set Membership: |
|

Ccdc37
Genomic Map