Assay Details
| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749786.3 | XP_003749834.1 | 1 - 2 | 269 | 87 | |
| XM_006236797.2 | XP_006236859.1 | 1 - 2 | 296 | 87 | ||
| XM_008763097.1 | XP_008761319.1 | 1 - 2 | 285 | 87 | ||
More Information
Set Membership: |
|

Pradc1
Genomic Map