Assay Details
| Gene Symbol: Podxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106607.1 | NP_001100077.1 | 2 - 3 | 345 | 109 | |
| GenBank mRNA | BC161970.1 | - | 2 - 3 | 387 | 9028963 | 109 |
More Information
Updated Mapping Information
Set Membership: |
|

Podxl2
Genomic Map