Assay Details
| Gene Symbol: Podxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106607.1 | NP_001100077.1 | 4 - 5 | 1216 | 148 | |
| GenBank mRNA | BC161970.1 | - | 4 - 5 | 1258 | 9028963 | 148 |
More Information
Updated Mapping Information
Set Membership: |
|

Podxl2
Genomic Map