Assay Details
| Gene Symbol: Nfu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106606.2 | NP_001100076.2 | 7 - 8 | 711 | 127 | |
| XM_008763063.1 | XP_008761285.1 | 7 - 8 | 944 | 127 | ||
| GenBank mRNA | FM064033.1 | - | 6 - 7 | 537 | 127 | |
| FM092780.1 | - | 1 - 2 | 127 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nfu1
Genomic Map