Assay Details
| Gene Symbol: Arpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106615.2 | NP_001100085.2 | 2 - 3 | 148 | 101 | |
| GenBank mRNA | BC167049.1 | - | 2 - 3 | 136 | 7442696 | 101 |
| CK478610.1 | - | 2 - 3 | 148 | 101 | ||
| FQ228740.1 | - | 2 - 3 | 132 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arpc4
Genomic Map