Assay Details
| Gene Symbol: Serpinb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153733.2 | NP_714955.2 | 3 - 4 | 291 | 103 | |
| GenBank mRNA | BC061735.1 | - | 3 - 4 | 291 | 5598648 | 103 |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb10
Genomic Map