Assay Details
| Gene Symbol: Serpinb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153733.2 | NP_714955.2 | 5 - 6 | 548 | 98 | |
| GenBank mRNA | BC061735.1 | - | 5 - 6 | 548 | 5598648 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb10
Genomic Map